Anti-SRPK1 Mouse Monoclonal Antibody [clone: 6G9]

Supplier: Novus Biologicals
H00006732-M03
103310-756EA 798.46 USD
103310-756
Anti-SRPK1 Mouse Monoclonal Antibody [clone: 6G9]
Antibodies
The SRPK1 Antibody (6G9) from Novus Biologicals is a mouse monoclonal antibody to SRPK1. This antibody reacts with human. The SRPK1 Antibody (6G9) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SRPK1
Clonality: Monoclonal
Clone: 6G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR