Anti-SRGAP2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-88830
103276-080EA 734.58 USD
103276-080
Anti-SRGAP2 Rabbit Polyclonal Antibody
Antibodies
The SRGAP2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SRGAP2. This antibody reacts with human. The SRGAP2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SRGAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR