Anti-SREBP2 Mouse Monoclonal Antibody [clone: 1D2]

Supplier: Novus Biologicals
NBP1-54446SS
103307-232EA 208 USD
103307-232
Anti-SREBP2 Mouse Monoclonal Antibody [clone: 1D2]
Antibodies
The SREBP2 Antibody (1D2) from Novus Biologicals is a mouse monoclonal antibody to SREBP2. This antibody reacts with human, chicken. The SREBP2 Antibody (1D2) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Gel Super Shift Assays.

Type: Primary
Antigen: SREBP2
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR