Anti-SRD5A1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-31909
103287-172EA 734.58 USD
103287-172
Anti-SRD5A1 Rabbit Polyclonal Antibody
Antibodies
The SRD5A1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SRD5A1. This antibody reacts with human. The SRD5A1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SRD5A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR