Anti-SRCAP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-84063
103268-332EA 734.58 USD
103268-332
Anti-SRCAP Rabbit Polyclonal Antibody
Antibodies
The SRCAP Antibody from Novus Biologicals is a rabbit polyclonal antibody to SRCAP. This antibody reacts with human. The SRCAP Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SRCAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR