Anti-SR-BI Goat Polyclonal Antibody (Alexa Fluor® 700)

Supplier: Novus Biologicals
NB400-131AF700
103409-178EA 323 USD
103409-178
Anti-SR-BI Goat Polyclonal Antibody (Alexa Fluor® 700)
Antibodies
The SR-BI Antibody [Alexa Fluor« 700] from Novus Biologicals is a goat polyclonal antibody to SR-BI. This antibody reacts with human, mouse, rat, hamster, mustelid, squirrel. The SR-BI Antibody [Alexa Fluor« 700] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: SR-BI
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 700
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster, Mustelid, Squirrel
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR