Anti-SR-BI Goat Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Novus Biologicals
NB400-131AF647
103407-894EA 323 USD
103407-894
Anti-SR-BI Goat Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
The SR-BI Antibody [Alexa Fluor« 647] from Novus Biologicals is a goat polyclonal antibody to SR-BI. This antibody reacts with human, mouse, rat, hamster, mustelid, squirrel. The SR-BI Antibody [Alexa Fluor« 647] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: SR-BI
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster, Mustelid, Squirrel
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR