Anti-SR1 Mouse Monoclonal Antibody [clone: 1E12.]

H00006717-M01
103310-718EA 583.08 USD
103310-718
Anti-SR1 Mouse Monoclonal Antibody [clone: 1E12.]
The SR1 Antibody (1E12.) from Novus Biologicals is a mouse monoclonal antibody to SR1. This antibody reacts with human, mouse, rat. The SR1 Antibody (1E12.) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SR1
Clonality: Monoclonal
Clone: 1E12.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR