Anti-SPT3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-87007
103273-982EA 734.58 USD
103273-982
Anti-SPT3 Rabbit Polyclonal Antibody
Antibodies
The SPT3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPT3. This antibody reacts with human. The SPT3 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR