Anti-SPSB2 Mouse Monoclonal Antibody [clone: 1E6]

Supplier: Novus Biologicals
H00084727-M01
103345-378EA 798.46 USD
103345-378
Anti-SPSB2 Mouse Monoclonal Antibody [clone: 1E6]
Antibodies
The SPSB2 Antibody (1E6) from Novus Biologicals is a mouse monoclonal antibody to SPSB2. This antibody reacts with human. The SPSB2 Antibody (1E6) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPSB2
Clonality: Monoclonal
Clone: 1000000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR