Anti-SPRYD7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-92443
103284-356EA 734.58 USD
103284-356
Anti-SPRYD7 Rabbit Polyclonal Antibody
Antibodies
The SPRYD7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPRYD7. This antibody reacts with human, mouse, rat. The SPRYD7 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPRYD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR