Anti-SPRY2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-13158
102713-256EA 777.05 USD
102713-256
Anti-SPRY2 Rabbit Polyclonal Antibody
Antibodies
The SPRY2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPRY2. This antibody reacts with human. The SPRY2 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPRY2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR