Anti-SPRED2 Mouse Monoclonal Antibody [clone: 6G8]

Supplier: Novus Biologicals
H00200734-M01
103347-694EA 798.46 USD
103347-694
Anti-SPRED2 Mouse Monoclonal Antibody [clone: 6G8]
Antibodies
The SPRED2 Antibody (6G8) from Novus Biologicals is a mouse monoclonal antibody to SPRED2. This antibody reacts with human. The SPRED2 Antibody (6G8) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SPRED2
Clonality: Monoclonal
Clone: 6G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR