Anti-SPR Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-87060
103269-856EA 734.58 USD
103269-856
Anti-SPR Rabbit Polyclonal Antibody
Antibodies
The SPR Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPR. This antibody reacts with human. The SPR Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR