Anti-SPPL2a Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-87512
103270-652EA 734.58 USD
103270-652
Anti-SPPL2a Rabbit Polyclonal Antibody
Antibodies
The SPPL2a Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPPL2a. This antibody reacts with human. The SPPL2a Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPPL2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR