Anti-SPOCK3 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00050859-B01P
103343-520EA 399 USD
103343-520
Anti-SPOCK3 Mouse Polyclonal Antibody
Antibodies
The Testican 3 / SPOCK3 Antibody from Novus Biologicals is a mouse polyclonal antibody to Testican 3 / SPOCK3. This antibody reacts with human. The Testican 3 / SPOCK3 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SPOCK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR