Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPOCK1 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00006695-B02P
The Testican 1 / SPOCK1 Antibody from Novus Biologicals is a mouse polyclonal antibody to Testican 1 / SPOCK1. This antibody reacts with human. The Testican 1 / SPOCK1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SPOCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.05 mg H00006695-B02P 103310-650 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol SPOCK1
Antigen Name Testican 1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Mouse
Isotype IgG
Western Blot Yes
Antigen Synonyms FLJ37170, sparc/osteonectin, Protein SPOCK, SPOCKcwcv and kazal-like domains proteoglycan (testican), cwcv and kazal-like domains proteoglycan (testican) 1
Gene ID 6695
Immunogen SPOCK1 (AAH30691.1, 1 a.a. - 439 a.a.) full-length human protein. MPAIAVLAAAAAAWCFLQVESRHLDALAGGAGPNHGNFLDNDQWLSTVSQYDRDKYWNRFRDDDYFRNWNPNKPFDKALDPSKDPCLKVKCSPHKVCVTQDYQTALCVSRKHLLPRQKKGNVAQKHWVGPSNLVKCKPCPVAQSAMVCGSDGHSYTSKCKLEFHACSTGKSLATLCDGPCPCLPEPEPPKHKAERSACTDKELRNLASRLKDWFGALHEDANRVIKPTSSNTAQGRFDTSILPICKDSLGWMFNKLDMNYDLLLDPSEINAIYLDKYEPCIKPLFNSCDSFKDGKLSNNEWCYCFQKPGGLPCQNEMNRIQKLSKGKSLLGAFIPRCNEEGYYKATQCHGSTGQCWCVDKYGNELAGSRKQGAVSCEEEQETSGDFGSGGSVVLLDDLEYERELGPKDKEGKLRVHTRAVTEDDEDEDDDKEDEVGYIW
Purification Protein A purified
Storage Buffer PBS (pH 7.4)
Storage Temperature Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty