Anti-SPINK5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-90510
103280-380EA 734.58 USD
103280-380
Anti-SPINK5 Rabbit Polyclonal Antibody
Antibodies
The SPINK5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPINK5. This antibody reacts with human. The SPINK5 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPINK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR