Anti-SPINK1 Mouse Monoclonal Antibody [clone: 4D4]

Supplier: Novus Biologicals
H00006690-M01
103310-638EA 798.46 USD
103310-638
Anti-SPINK1 Mouse Monoclonal Antibody [clone: 4D4]
Antibodies
The SPINK1 Antibody (4D4) from Novus Biologicals is a mouse monoclonal antibody to SPINK1. This antibody reacts with human. The SPINK1 Antibody (4D4) has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPINK1
Clonality: Monoclonal
Clone: 4D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR