Anti-SPHAR Goat Polyclonal Antibody

Supplier: Novus Biologicals
NB300-985
103295-204EA 303 USD
103295-204
Anti-SPHAR Goat Polyclonal Antibody
Antibodies
The SPHAR Antibody from Novus Biologicals is a goat polyclonal antibody to SPHAR. This antibody reacts with human. The SPHAR Antibody has been validated for the following applications: Peptide ELISA.

Type: Primary
Antigen: SPHAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR