Anti-SPG8 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20471
102711-812EA 635.51 USD
102711-812
Anti-SPG8 Rabbit Polyclonal Antibody
Antibodies
The SPG8 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPG8. This antibody reacts with human. The SPG8 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPG8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR