Anti-SPG3A Mouse Monoclonal Antibody [clone: 1B11]

Supplier: Novus Biologicals
H00051062-M10
103343-622EA 798.46 USD
103343-622
Anti-SPG3A Mouse Monoclonal Antibody [clone: 1B11]
Antibodies
The SPG3A Antibody (1B11) from Novus Biologicals is a mouse monoclonal antibody to SPG3A. This antibody reacts with human, rat. The SPG3A Antibody (1B11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SPG3A
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR