Anti-SPEN Goat Polyclonal Antibody

Supplier: Novus Biologicals
NB100-2804
103357-644EA 299 USD
103357-644
Anti-SPEN Goat Polyclonal Antibody
Antibodies
The SPEN Antibody from Novus Biologicals is a goat polyclonal antibody to SPEN. This antibody reacts with human. The SPEN Antibody has been validated for the following applications: Peptide ELISA.

Type: Primary
Antigen: SPEN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR