Anti-Speedy/Ringo Rabbit Polyclonal Antibody (DyLight 405)

Supplier: Novus Biologicals
NB100-2521V
103369-132EA 614.15 USD
103369-132
Anti-Speedy/Ringo Rabbit Polyclonal Antibody (DyLight 405)
Antibodies
The Speedy / Ringo Antibody [DyLight 405] from Novus Biologicals is a rabbit polyclonal antibody to Speedy / Ringo. This antibody reacts with human, mouse, rat, sheep. The Speedy / Ringo Antibody [DyLight 405] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Speedy/Ringo
Clonality: Polyclonal
Clone:
Conjugation: DyLight 405
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR