Anti-Speedy/Ringo Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Novus Biologicals
NB100-2521F
103369-130EA 614.15 USD
103369-130
Anti-Speedy/Ringo Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
The Speedy / Ringo Antibody [FITC] from Novus Biologicals is a rabbit polyclonal antibody to Speedy / Ringo. This antibody reacts with human, mouse, rat, sheep. The Speedy / Ringo Antibody [FITC] has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Speedy/Ringo
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR