Anti-SP-D Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-33424
103282-920EA 734.58 USD
103282-920
Anti-SP-D Rabbit Polyclonal Antibody
Antibodies
The SP-D Antibody from Novus Biologicals is a rabbit polyclonal antibody to SP-D. This antibody reacts with human. The SP-D Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SP-D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR