Anti-SP-D Mouse Monoclonal Antibody [clone: 2C10]

H00006441-M03
103310-064EA 553.85 USD
103310-064
Anti-SP-D Mouse Monoclonal Antibody [clone: 2C10]
The SP-D Antibody (2C10) from Novus Biologicals is a mouse monoclonal antibody to SP-D. This antibody reacts with human. The SP-D Antibody (2C10) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SP-D
Clonality: Monoclonal
Clone: 2C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR