Anti-SP-D Mouse Monoclonal Antibody [clone: 3E6.]

H00006441-M01
103310-060EA 553.85 USD
103310-060
Anti-SP-D Mouse Monoclonal Antibody [clone: 3E6.]
The SP-D Antibody (3E6.) from Novus Biologicals is a mouse monoclonal antibody to SP-D. This antibody reacts with human. The SP-D Antibody (3E6.) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SP-D
Clonality: Monoclonal
Clone: 3E6.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR