Anti-SP-B Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00006439-B01P
103310-048EA 798.46 USD
103310-048
Anti-SP-B Mouse Polyclonal Antibody
Antibodies
The SP-B / Surfactant Protein B Antibody from Novus Biologicals is a mouse polyclonal antibody to SP-B / Surfactant Protein B. This antibody reacts with human. The SP-B / Surfactant Protein B Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SP-B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR