Anti-SPATA4 Mouse Monoclonal Antibody [clone: 4A2]

Supplier: Novus Biologicals
H00132851-M03
103346-840EA 798.46 USD
103346-840
Anti-SPATA4 Mouse Monoclonal Antibody [clone: 4A2]
Antibodies
The SPATA4 Antibody (4A2) from Novus Biologicals is a mouse monoclonal antibody to SPATA4. This antibody reacts with human. The SPATA4 Antibody (4A2) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SPATA4
Clonality: Monoclonal
Clone: 4A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR