Anti-SPATA2 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00009825-B01P
103337-654EA 399 USD
103337-654
Anti-SPATA2 Mouse Polyclonal Antibody
Antibodies
The SPATA2 Antibody from Novus Biologicals is a mouse polyclonal antibody to SPATA2. This antibody reacts with human. The SPATA2 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SPATA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR