Anti-SPATA2 Mouse Monoclonal Antibody [clone: 1F1]

Supplier: Novus Biologicals
H00009825-M01
103337-658EA 798.46 USD
103337-658
Anti-SPATA2 Mouse Monoclonal Antibody [clone: 1F1]
Antibodies
The SPATA2 Antibody (1F1) from Novus Biologicals is a mouse monoclonal antibody to SPATA2. This antibody reacts with human. The SPATA2 Antibody (1F1) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SPATA2
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR