Anti-SPATA19 Chicken Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-41149
103405-794EA 564.74 USD
103405-794
Anti-SPATA19 Chicken Polyclonal Antibody
Antibodies
The SPATA19 Antibody from Novus Biologicals is a chicken polyclonal antibody to SPATA19. This antibody reacts with human, mouse, rat. The SPATA19 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SPATA19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR