Anti-SPATA16 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20466
102711-802EA 635.51 USD
102711-802
Anti-SPATA16 Rabbit Polyclonal Antibody
Antibodies
The SPATA16 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPATA16. This antibody reacts with human. The SPATA16 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPATA16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR