Anti-SPATA13 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-90849
103278-820EA 734.58 USD
103278-820
Anti-SPATA13 Rabbit Polyclonal Antibody
Antibodies
The SPATA13 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPATA13. This antibody reacts with human. The SPATA13 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SPATA13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR