Anti-Spastin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-90230
103280-224EA 734.58 USD
103280-224
Anti-Spastin Rabbit Polyclonal Antibody
Antibodies
The Spastin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Spastin. This antibody reacts with human, rat. The Spastin Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Spastin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR