Anti-SPARC Rabbit Polyclonal Antibody

H00006678-D01P
103310-618EA 583.08 USD
103310-618
Anti-SPARC Rabbit Polyclonal Antibody
The SPARC Antibody from Novus Biologicals is a rabbit polyclonal antibody to SPARC. This antibody reacts with human. The SPARC Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SPARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR