Anti-Sp3 Mouse Monoclonal Antibody [clone: 2E5.]

Supplier: Novus Biologicals
H00006670-M07
103310-584EA 798.46 USD
103310-584
Anti-Sp3 Mouse Monoclonal Antibody [clone: 2E5.]
Antibodies
The Sp3 Antibody (2E5.) from Novus Biologicals is a mouse monoclonal antibody to Sp3. This antibody reacts with human. The Sp3 Antibody (2E5.) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Sp3
Clonality: Monoclonal
Clone: 2E5.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR