Anti-SP100 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-86060
103272-472EA 734.58 USD
103272-472
Anti-SP100 Rabbit Polyclonal Antibody
Antibodies
The SP100 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SP100. This antibody reacts with human. The SP100 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Micro.

Type: Primary
Antigen: SP100
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR