Anti-SP100 Mouse Monoclonal Antibody [clone: 1G6]

Supplier: Novus Biologicals
H00006672-M02
103310-606EA 798.46 USD
103310-606
Anti-SP100 Mouse Monoclonal Antibody [clone: 1G6]
Antibodies
The SP100 Antibody (1G6) from Novus Biologicals is a mouse monoclonal antibody to SP100. This antibody reacts with human. The SP100 Antibody (1G6) has been validated for the following applications: Western Blot, ELISA, RNA Inhibition.

Type: Primary
Antigen: SP100
Clonality: Monoclonal
Clone: 1G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR