Anti-SP100 Mouse Monoclonal Antibody [clone: 2E2.]

Supplier: Novus Biologicals
H00006672-M03
103310-608EA 798.46 USD
103310-608
Anti-SP100 Mouse Monoclonal Antibody [clone: 2E2.]
Antibodies
The SP100 Antibody (2E2.) from Novus Biologicals is a mouse monoclonal antibody to SP100. This antibody reacts with human. The SP100 Antibody (2E2.) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SP100
Clonality: Monoclonal
Clone: 2E2.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR