Anti-SP1 Mouse Monoclonal Antibody [clone: 2G5]

H00006667-M08
103310-572EA 553.85 USD
103310-572
Anti-SP1 Mouse Monoclonal Antibody [clone: 2G5]
The SP1 Antibody (2G5) from Novus Biologicals is a mouse monoclonal antibody to SP1. This antibody reacts with human, mouse. The SP1 Antibody (2G5) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SP1
Clonality: Monoclonal
Clone: 2G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR