Anti-SOX8 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-68807
103407-876EA 299 USD
103407-876
Anti-SOX8 Goat Polyclonal Antibody
Antibodies
The SOX8 Antibody from Novus Biologicals is a goat polyclonal antibody to SOX8. This antibody reacts with human, mouse, rat. The SOX8 Antibody has been validated for the following applications: Peptide ELISA.

Type: Primary
Antigen: SOX8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-EGRRPVDSGRQN
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR