Anti-SOX8 Mouse Monoclonal Antibody [clone: 8D8]

Supplier: Novus Biologicals
H00030812-M02
103343-304EA 798.46 USD
103343-304
Anti-SOX8 Mouse Monoclonal Antibody [clone: 8D8]
Antibodies
The SOX8 Antibody (8D8) from Novus Biologicals is a mouse monoclonal antibody to SOX8. This antibody reacts with human. The SOX8 Antibody (8D8) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SOX8
Clonality: Monoclonal
Clone: 8D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR