Anti-SOX30 Mouse Monoclonal Antibody [clone: 1A11]

Supplier: Novus Biologicals
H00011063-M01
103340-062EA 798.46 USD
103340-062
Anti-SOX30 Mouse Monoclonal Antibody [clone: 1A11]
Antibodies
The SOX30 Antibody (1A11) from Novus Biologicals is a mouse monoclonal antibody to SOX30. This antibody reacts with human. The SOX30 Antibody (1A11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SOX30
Clonality: Monoclonal
Clone: 1A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR