Anti-SOX22 Mouse Monoclonal Antibody [clone: 2A6]

Supplier: Novus Biologicals
H00006666-M06
103310-558EA 798.46 USD
103310-558
Anti-SOX22 Mouse Monoclonal Antibody [clone: 2A6]
Antibodies
The SOX22 Antibody (2A6) from Novus Biologicals is a mouse monoclonal antibody to SOX22. This antibody reacts with human. The SOX22 Antibody (2A6) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SOX22
Clonality: Monoclonal
Clone: 2A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR