Anti-SOX15 Mouse Monoclonal Antibody [clone: 1B3]

Supplier: Novus Biologicals
H00006665-M12
103310-552EA 798.46 USD
103310-552
Anti-SOX15 Mouse Monoclonal Antibody [clone: 1B3]
Antibodies
The SOX15 Antibody (1B3) from Novus Biologicals is a mouse monoclonal antibody to SOX15. This antibody reacts with human. The SOX15 Antibody (1B3) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SOX15
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR