Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SOX10 Goat Polyclonal Antibody (DyLight 550)

Supplier: Novus Biologicals
NBP2-52744R
The SOX10 Antibody [DyLight 550] from Novus Biologicals is a goat polyclonal antibody to SOX10. This antibody reacts with human. The SOX10 Antibody [DyLight 550] has been validated for the following applications: Western Blot, Immunohistochemistry.

Type: Primary
Antigen: SOX10
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP2-52744R 103409-170 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol SOX10
Reactivity Human
Conjugation DyLight® 550
Clonality Polyclonal
Antibody Type Primary
Host Goat
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms PCWH, MGC15649, SRY (sex determining region Y)-box 10, WS4C, transcription factor SOX-10, WS4mouse, SRY-related HMG-box gene 10, human homolog of
Gene ID 6663
Immunogen <EM>E. coli</EM>-derived recombinant human SOX10, Met1-Ala118, Accession # P56693
Purification Antigen Affinity-purified
Storage Buffer 50mM Sodium Borate
Storage Temperature Store at 4C in the dark.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty