Anti-SOX10 Mouse Polyclonal Antibody

H00006663-B01P
103310-546EA 553.85 USD
103310-546
Anti-SOX10 Mouse Polyclonal Antibody
The SOX10 Antibody from Novus Biologicals is a mouse polyclonal antibody to SOX10. This antibody reacts with human. The SOX10 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SOX10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR