Anti-SORBS1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-86641
103273-836EA 734.58 USD
103273-836
Anti-SORBS1 Rabbit Polyclonal Antibody
Antibodies
The SORBS1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SORBS1. This antibody reacts with human. The SORBS1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.

Type: Primary
Antigen: SORBS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR